Läs mer
Peramivir, TRC
Produktkod. 30409291
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30409291 1 mg
1 options
Produktkod. 30409291 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2855001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Peramivir
CAS 330600-85-6
Molekylformel C15 H28 N4 O4
Kvantitet 1 mg
Synonym Cyclopentanecarboxylic acid','3-[(1S)-1-(acetylamino)-2-ethylbutyl]-4-[(aminoiminomethyl)amino]-2-hydroxy-','(1S,2S,3R,4R)-, (1S,2S,3R,4R)-3-[(1S)-1-(Acetylamino)-2-ethylbutyl]-4-[(aminoiminomethyl)amino]-2-hydroxycyclopentanecarboxylic acid, (1S,2S,3R,4R)-3-((1S)-1-Acetamido-2-ethylbutyl)-4-(diaminomethylideneamino)-2-hydroxycyclopentane-1-carboxylic acid, (1S,2S,3S,4R)-3-((1S)-1-Acetamido-2-ethylbutyl)-4-(diaminomethylideneamino)-2-hydroxycyclopentane-1-carboxylic acid, BCX 1812, Peramivir, RWJ 270201, Rapiacta, Rapivab
InChI formel InChI=1S/C15H28N4O4/c1-4-8(5-2)12(18-7(3)20)11-10(19-15(16)17)6-9(13(11)21)14(22)23/h8-13,21H,4-6H2,1-3H3,(H,18,20)(H,22,23)(H4,16,17,19)/t9-,10+,11+,12-,13+/m0/s1
LEDER CCC(CC)[C@H](NC(=O)C)[C@@H]1[C@H](O)[C@H](C[C@H]1NC(=N)N)C(=O)O
IUPAC-namn (1S,2S,3R,4R)-3-[(1S)-1-acetamido-2-ethylbutyl]-4-carbamimidamido-2-hydroxycyclopentane-1-carboxylic acid
Molekylvikt (g/mol) 328.41
Formel vikt 328.2111