Läs mer
Perazine-d8 Dihydrochloride Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Perazine-d8 Dihydrochloride Salt |
| CAS | 1246815-57-5 |
| Molekylformel | C20H19D8Cl2N3S |
| Kvantitet | 10 mg |
| Synonym | 10-[3-(4-Methyl-1-piperazinyl)propyl]-10H-phenothiazine-d8 Dihydrochloride Salt, 10-[3-(4-Methyl-1-piperazinyl)propyl]phenothiazine-d8 Dihydrochloride Salt, 10-[3-(4-Methylpiperazinyl)propyl]phenothiazine-d8 Dihydrochloride Salt, NSC 638694-d8 Dihydrochloride Salt, P 725-d8 Dihydrochloride Salt, Perazine-d8 Dihydrochloride Salt, Pernazine-d8 Dihydrochloride Salt, Psytomin-d8 Dihydrochloride Salt, Taxilan-d8 Dihydrochloride Salt |
| InChI formel | InChI=1S/C20H25N3S/c1-21-13-15-22(16-14-21)11-6-12-23-17-7-2-4-9-19(17)24-20-10-5-3-8-18(20)23/h2-5,7-10H,6,11-16H2,1H3/i13D2,14D2,15D2,16D2 |
| LEDER | CN(C([2H])([2H])C1([2H])[2H])C([2H])([2H])C([2H])([2H])N1CCCN2C3=C(C=CC=C3)SC4=CC=CC=C42 |
| Molekylvikt (g/mol) | 420.47 |