Läs mer
Perazine Dimalonate, LoGiCal
Specifikationer
Specifikationer
| Kemiskt namn eller material | Perazine Dimalonate |
| CAS | 14777-25-4 |
| Analyt- eller komponentnamn | Perazine Dimalonate |
| Frakt skick | Room Temperature |
| Molekylformel | C20 H25 N3 S . 2 C3 H4 O4 |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Perazine Dimalonate, Propanedioic acid compd. with 10-[3-(4-methyl-1-piperazinyl)propyl]-10H-phenothiazine (2:1), Malonic acid compd. with 10-[3-(4-methyl-1-piperazinyl)propyl]phenothiazine (2:1), 10-[3-(4-Methyl-1-piperazinyl)propyl]-10H-phenothiazine propanedioate (1:2), 10-[3-(4-Methyl-1-piperazinyl)propyl]phenothiazine malonate (1:2), Perazine acid malonate, Perazine dimalonate, Perazine malonate, Prochlorperazine Maleate Impurity B as Dimalonate, Prochlorperazine Impurity B as Dimalonate |
| InChI formel | InChI=1S/C20H25N3S.2C3H4O4/c1-21-13-15-22(16-14-21)11-6-12-23-17-7-2-4-9-19(17)24-20-10-5-3-8-18(20)23;2*4-2(5)1-3(6)7/h2-5,7-10H,6,11-16H2,1H3;2*1H2,(H,4,5)(H,6,7) |
| Lösningstyp | Neat |
| LEDER | CN1CCN(CCCN2c3ccccc3Sc4ccccc24)CC1.OC(=O)CC(=O)O.OC(=O)CC(=O)O |
| Visa mer |