missing translation for 'onlineSavingsMsg'
Läs mer

Perazine Dimalonate, Mikromol™

Produktkod. 17901416
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17901416 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17901416 Leverantör Mikromol Leverantörsnummer MM0157.000250

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Perazine Dimalonate
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Perazine Dimalonate
CAS 14777-25-4
Analyt- eller komponentnamn Perazine Dimalonate
Frakt skick Room Temperature
Molekylformel C20 H25 N3 S . 2 C3 H4 O4
Certifikat och efterlevnad ISO 17034
Synonym Perazine Dimalonate, Propanedioic acid compd. with 10-[3-(4-methyl-1-piperazinyl)propyl]-10H-phenothiazine (2:1), Malonic acid compd. with 10-[3-(4-methyl-1-piperazinyl)propyl]phenothiazine (2:1), 10-[3-(4-Methyl-1-piperazinyl)propyl]-10H-phenothiazine propanedioate (1:2), 10-[3-(4-Methyl-1-piperazinyl)propyl]phenothiazine malonate (1:2), Perazine acid malonate, Perazine dimalonate, Perazine malonate, Prochlorperazine Maleate Impurity B as Dimalonate, Prochlorperazine Impurity B as Dimalonate
InChI formel InChI=1S/C20H25N3S.2C3H4O4/c1-21-13-15-22(16-14-21)11-6-12-23-17-7-2-4-9-19(17)24-20-10-5-3-8-18(20)23;2*4-2(5)1-3(6)7/h2-5,7-10H,6,11-16H2,1H3;2*1H2,(H,4,5)(H,6,7)
LEDER CN1CCN(CCCN2c3ccccc3Sc4ccccc24)CC1.OC(=O)CC(=O)O.OC(=O)CC(=O)O
IUPAC-namn 10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine;propanedioic acid
Molekylvikt (g/mol) 547.62
Kvantitet 250 mg
Formel vikt 547.1988 g/mol
Visa mer Visa mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.