Läs mer
Perazine Sulphoxide, Mikromol™
Produktkod. 30049469
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049469 25 mg
17983166 100 mg
30461972 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30049469 Leverantör Mikromol Leverantörsnummer MM0157.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Perazine Sulphoxide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perazine Sulphoxide
CAS 20627-44-5
Analyt- eller komponentnamn Perazine Sulphoxide
Frakt skick Room Temperature
Molekylformel C20 H25 N3 O S
Certifikat och efterlevnad ISO 17025
Synonym Perazine Sulphoxide
InChI formel InChI=1S/C20H25N3OS/c1-21-13-15-22(16-14-21)11-6-12-23-17-7-2-4-9-19(17)25(24)20-10-5-3-8-18(20)23/h2-5,7-10H,6,11-16H2,1H3
LEDER CN1CCN(CCCN2c3ccccc3S(=O)c4ccccc24)CC1
IUPAC-namn 10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine 5-oxide
Molekylvikt (g/mol) 355.5
Kvantitet 25 mg
Formel vikt 355.1718 g/mol