missing translation for 'onlineSavingsMsg'
Läs mer

Perazine Sulphoxide, Mikromol™

Produktkod. 30461972
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461972 4 x 25 mg
30049469 25 mg
17983166 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30461972 Supplier Mikromol Supplier No. MM0157.0100254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Perazine Sulphoxide, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Perazine Sulphoxide
CAS 20627-44-5
Analyt- eller komponentnamn Perazine Sulphoxide
Frakt skick Room Temperature
Molekylformel C20 H25 N3 O S
Certifikat och efterlevnad ISO 17025
Synonym Perazine Sulphoxide
InChI formel InChI=1S/C20H25N3OS/c1-21-13-15-22(16-14-21)11-6-12-23-17-7-2-4-9-19(17)25(24)20-10-5-3-8-18(20)23/h2-5,7-10H,6,11-16H2,1H3
Lösningstyp Neat
LEDER CN1CCN(CCCN2c3ccccc3S(=O)c4ccccc24)CC1
IUPAC-namn 10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine 5-oxide
Molekylvikt (g/mol) 355.5
Kvantitet 4 x 25 mg
Formel vikt 355.1718 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.