missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid, 0.1M, Chem Lab™
Click to view available options
Quantity:
1 L
Grade:
Laboratory
Reagent Acc. Ph. Eur. 9(4.2.2)
Specifications
pH | <2 |
Color | Colorless |
Density | 1.05g/mL |
Odor | Acetic Acid |
Packaging | 1L |
Quantity | 1 L |
Physical Form | Liquid |
Grade | Laboratory |
Safety and Handling
Product Identifier
- Perchloric acid 0.1 mol/l in Acetic acid 100%
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : perchloric acid, acetic acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'