Läs mer
Perfluoro-N-methylmorpholine, TRC
Produktkod. 30410452
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30410452 250 mg
1 options
Produktkod. 30410452 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP010025250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluoro-N-methylmorpholine
CAS 382-28-5
Molekylformel C5 F11 N O
Kvantitet 250 mg
Synonym Perfluoro-N-methylmorpholine, Morpholine','2,2,3,3,5,5,6,6-octafluoro-4-(trifluoromethyl)- (6CI','7CI','8CI','9CI','ACI), 2,2,3,3,5,5,6,6-Octafluoro-4-(trifluoromethyl)morpholine (ACI), 4-(Trifluoromethyl)perfluoromorpholine, Fluorinert PF 5052, L 12422, Perfluoro(4-methylmorpholine), Perfluoro-N-methylmorpholine, PF 5052, PFC 5052
InChI formel InChI=1S/C5F11NO/c6-1(7)3(10,11)18-4(12,13)2(8,9)17(1)5(14,15)16
LEDER FC(F)(F)N1C(F)(F)C(F)(F)OC(F)(F)C1(F)F
IUPAC-namn 2,2,3,3,5,5,6,6-octafluoro-4-(trifluoromethyl)morpholine
Molekylvikt (g/mol) 299.042
Formel vikt 298.98