Läs mer
Perfluoro-n-pentane, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dodecafluoropentane |
| CAS | 678-26-2 |
| Anteckningar om renhetsgrad | GC |
| Molekylformel | C5 F12 |
| Kvantitet | 5 g |
| Synonym | Perfluoropentane, Dodecafluoropentane, Pentane','1,1,1,2,2,3,3,4,4,5,5,5-dodecafluoro-, Pentane','dodecafluoro- (6CI,7CI,8CI,9CI), 1,1,1,2,2,3,3,4,4,5,5,5-Dodecafluoropentane, Dodecafluoropentane, FC 4112, FC 87, Fluorinert FC 87, Fluorinert PF 5050, Flutec PP 50, NVX 108, PF 5050, PFC 41-12, Perflenapent, Perfluoro-n-pentane, Perfluoropentane, QW 7437, R 41(12), R-41121,1,1,2,2,3,3,4,4,5,5,5-Dodecafluoropentane, Dodecafluoropentane, FC 4112, FC 87, Fluorinert FC 87, Fluorinert PF 5050, Flutec PP 50, NVX 108, PF 5050, PFC 41-12, Perflenapent, Perfluoro-n-pentane, Perfluoropentane, QW 7437, R 41(12), R-4112 |
| InChI formel | InChI=1S/C5F12/c6-1(7,2(8,9)4(12,13)14)3(10,11)5(15,16)17 |
| LEDER | FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC-namn | 1,1,1,2,2,3,3,4,4,5,5,5-dodecafluoropentane |
| Molekylvikt (g/mol) | 288.034 |
| Visa mer |