Läs mer
Perfluoro-octylethanoic Acid, TRC
Produktkod. 17973795
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17973795 250 mg
17943645 100 mg
17984955 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17973795 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP286258250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27854-31-5
Molekylformel C10 H3 F17 O2
Molekylvikt (g/mol) 478.1
Synonym Decanoic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecanoic acid, 8:2 FTCA, 8:2 Fluorotelomer carboxylic acid, Perfluoro-octylethanoic acid, 2H,2H-Perfluorodecanoic acid
IUPAC-namn 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecanoic acid
LEDER OC(=O)CC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material 2H,2H-Perfluorodecanoic Acid
Kvantitet 250 mg
InChI formel InChI=1S/C10H3F17O2/c11-3(12,1-2(28)29)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)9(23,24)10(25,26)27/h1H2,(H,28,29)
Formel vikt 477.9862