Learn More
Perfluoro-tert-butanol, 99%
Product Code. 11480587
Change view
Click to view available options
Kvantitet:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
11480587 1 g 1g
11490587 5 g 5g
11400597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11480587 Supplier Thermo Scientific Alfa Aesar Supplier No. B22442.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 2378-02-1 | C4HF9O | 236.04 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2378-02-1
MDL-nummer MFCD00042092
Specifications
Kemiskt namn eller material Perfluoro-tert-butanol
Smältpunkt -17°C
Kokpunkt 44°C to 45°C
Odör Characteristic
Molekylformel C4HF9O
Linjär formel (CF3)3COH
Kvantitet 1 g
UN-nummer UN2810
Beilstein 1841640
Synonym perfluoro-tert-butanol, nonafluoro-tert-butanol, perfluoro-tert-butyl alcohol, perfluoro-t-butanol, nonafluoro-tert-butyl alcohol, 2-propanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl, nonafluor-terc.butanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl propan-2-ol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl-2-propanol, nonafluor-terc.butanol czech
InChI-nyckel XZNOAVNRSFURIR-UHFFFAOYSA-N
LEDER C(C(F)(F)F)(C(F)(F)F)(C(F)(F)F)O
IUPAC-namn 1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol
Molekylvikt (g/mol) 236.04
PubChem CID 16924
Formel vikt 236.04
Procent renhet 99%