Läs mer
Perfluorobutanesulfonic acid 13C3 (2,3,4-13C3) sodium 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Produktkod. 30319332
Change view
Kvantitet:
1 mL
Produktkod. Kvantitet
30319332 1 mL
1 options
Produktkod. 30319332 Leverantör Dr. Ehrenstorfer Leverantörsnummer DREA15986518MW50
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluorobutanesulfonic Acid 13C3 (2,3,4-13C3) Sodium
CAS 2708218-84-0
Lösningsmedel eller matris Methanol/Water
Molekylformel 13C3 C F9 O3 S . Na
Kvantitet 1 mL
Synonym 13C3-PFBSNa, Perfluorobutanesulfonic Acid 13C3 (2,3,4-13C3) Sodium, 1-Butane-2,3,4-13C3-sulfonic acid, 1,1,2,2,3,3,4,4,4-nonafluoro-, sodium salt (1:1) (ACI)
InChI formel InChI=1S/C4HF9O3S.Na/c5-1(6,3(9,10)11)2(7,8)4(12,13)17(14,15)16;/h(H,14,15,16);/q;+1/p-1/i1+1,2+1,3+1;
LEDER [Na+].[O-]S(=O)(=O)C(F)(F)[13C](F)(F)[13C](F)(F)[13C](F)(F)F
IUPAC-namn sodium;1,1,2,2,3,3,4,4,4-nonafluoro(2,3,4-13C3)butane-1-sulfonate
Molekylvikt (g/mol) 325.059
Formel vikt 324.942 g/mol