Läs mer
Perfluorobutylphosphonic Acid, TRC
Produktkod. 30422853
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30422853 50 mg
1 options
Produktkod. 30422853 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP28759550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluorobutylphosphonic Acid
CAS 52299-24-8
Anteckningar om renhetsgrad HPLC
Molekylformel C4H2F9O3P
Kvantitet 50 mg
Synonym P-(1,1,2,2,3,3,4,4,4-Nonafluorobutyl)phosphonic acid, 1,1,2,2,3,3,4,4,4-Nonafluorobutylphosphonic acid, Nonafluorobutylphosphonic acid
InChI formel InChI=1S/C4H2F9O3P/c5-1(6,3(9,10)11)2(7,8)4(12,13)17(14,15)16/h(H2,14,15,16)
LEDER OP(=O)(O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 1,1,2,2,3,3,4,4,4-nonafluorobutylphosphonic acid
Molekylvikt (g/mol) 300.02
Formel vikt 299.9598