Läs mer
Perfluorobutylsulphonamide, TRC
Produktkod. 30420741
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30420741 50 mg
1 options
Produktkod. 30420741 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP30335050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluorobutane Sulfonamide
CAS 30334-69-1
Anteckningar om renhetsgrad HPLC
Molekylformel C4 H2 F9 N O2 S
Kvantitet 50 mg
Synonym Perfluorobutane Sulfonamide, 1,1,2,2,3,3,4,4,4-Nonafluoro-1-butanesulfonamide, 1,1,2,2,3,3,4,4,4-Nonafluoro-n-butanesulfonamide, Nonafluorobutanesulfonamide, Perfluorobutanesulfonamide, n-Nonafluorobutanesulfonamide, FBSA, FBSA, Perfluorobutane Sulfonamide, 1,1,2,2,3,3,4,4,4-Nonafluoro-1-butanesulfonamide, 1,1,2,2,3,3,4,4,4-Nonafluoro-n-butanesulfonamide, Nonafluorobutanesulfonamide, Perfluorobutanesulfonamide, n-Nonafluorobutanesulfonamide, PFBSA
InChI formel InChI=1S/C4H2F9NO2S/c5-1(6,3(9,10)11)2(7,8)4(12,13)17(14,15)16/h(H2,14,15,16)
LEDER NS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonamide
Molekylvikt (g/mol) 299.11
Formel vikt 298.9663