Läs mer
Perfluorodecylphosphonic Acid, TRC
Produktkod. 30420685
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30420685 25 mg
1 options
Produktkod. 30420685 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP27062525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluorodecanephosphonic acid
CAS 52299-26-0
Anteckningar om renhetsgrad HPLC
Molekylformel C10 H2 F21 O3 P
Kvantitet 25 mg
Synonym Phosphonic acid','P-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heneicosafluorodecyl)-, Phosphonic acid','(heneicosafluorodecyl)- (9CI), P-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heneicosafluorodecyl)phosphonic acid, (Perfluorodecyl)phosphonic acid
InChI formel InChI=1S/C10H2F21O3P/c11-1(12,3(15,16)5(19,20)7(23,24)9(27,28)29)2(13,14)4(17,18)6(21,22)8(25,26)10(30,31)35(32,33)34/h(H2,32,33,34)
LEDER OP(=O)(O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-henicosafluorodecylphosphonic acid
Molekylvikt (g/mol) 600.06
Formel vikt 599.9406