Läs mer
Perfluoroheptyl Iodide, TRC
Produktkod. 30423214
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423214 1 g
30415412 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423214 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2863151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 335-58-0
Molekylformel C7F15I
Molekylvikt (g/mol) 495.96
Synonym 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-Pentadecafluoro-7-iodoheptane, Pentadecafluoro-1-iodoheptane, 1-Iodoperfluoroheptane, Pentadecafluoro-1-iodoheptane, Pentadecafluoroheptyl Iodide, Perfluoro-1-iodoheptane, Perfluoro-n-heptyl Iodide
IUPAC-namn 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-pentadecafluoro-7-iodoheptane
LEDER FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Specifikationer
Kemiskt namn eller material Perfluoroheptyl Iodide
Kvantitet 1 g
InChI formel InChI=1S/C7F15I/c8-1(9,2(10,11)4(14,15)6(18,19)20)3(12,13)5(16,17)7(21,22)23
Formel vikt 495.8805