Läs mer
Perfluorohexane Sulphonyl Fluoride (>80%), TRC
Produktkod. 30136124
Change view
Kvantitet:
1 g
2.5 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136124 5 g
30133842 2.5 g
30008647 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136124 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP9992505G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 423-50-7
Molekylformel C6F14O2S
Molekylvikt (g/mol) 402.11
Synonym 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluoro-1-hexanesulfonyl Fluoride, 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluorohexane-1-sulfonyl Fluoride, Perfluoro-1-hexanesulfonyl Fluoride, Perfluorohexanesulfonyl Fluoride, Perfluorohexylsulfonyl Fluoride, Tridecafluorohexanesulfonyl Fluoride
IUPAC-namn 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane-1-sulfonyl fluoride
LEDER FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)S(=O)(=O)F
Specifikationer
Kemiskt namn eller material Perfluorohexane Sulphonyl Fluoride
Kvantitet 5 g
InChI formel InChI=1S/C6F14O2S/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)23(20,21)22
Formel vikt 401.9395