Läs mer
Perfluorohexanoic acid 13C2 (1,2-13C2) 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Produktkod. 30317351
Change view
Kvantitet:
1 mL
Produktkod. Kvantitet
30317351 1 mL
1 options
Produktkod. 30317351 Leverantör Dr. Ehrenstorfer Leverantörsnummer DREA15986914MW50
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluorohexanoic Acid 13C2 (1,2-13C2)
CAS 960315-47-3
Lösningsmedel eller matris Methanol/Water
Molekylformel 13C2 C4 H F11 O2
Kvantitet 1 mL
Synonym Perfluorohexanoic Acid 13C2 (1,2-13C2), Hexanoic-1,2-13C2 acid, 2,2,3,3,4,4,5,5,6,6,6-undecafluoro- (ACI), 2,2,3,3,4,4,5,5,6,6,6-Undecafluorohexanoic-1,2-13C2 acid (ACI), Perfluoro-n-[1,2 13C2]hexanoic acid
InChI formel InChI=1S/C6HF11O2/c7-2(8,1(18)19)3(9,10)4(11,12)5(13,14)6(15,16)17/h(H,18,19)/i1+1,2+1
LEDER O[13C](=O)[13C](F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 2,2,3,3,4,4,5,5,6,6,6-undecafluoro(1,2-^{13}C_{2})hexanoic acid
Molekylvikt (g/mol) 316.039
Formel vikt 315.987 g/mol