Läs mer
Perfluorohexanoic acid 13C5 (1,2,3,4,6-13C5) 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Produktkod. 30326162
Change view
Kvantitet:
1 mL
Produktkod. Kvantitet
30326162 1 mL
1 options
Produktkod. 30326162 Leverantör Dr. Ehrenstorfer Leverantörsnummer DREA15986911MW50
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluorohexanoic Acid 13C5 (1,2,3,4,6-13C5)
CAS 2328024-54-8
Lösningsmedel eller matris Methanol/Water
Molekylformel 13C5 C H F11 O2
Kvantitet 1 mL
Synonym 13C5-PFHxA, Perfluorohexanoic Acid 13C5 (1,2,3,4,6-13C5), Perfluorohexanoic acid 13C5 (1,2,3,4,6-13C5), Perfluoro-n-[1,2,3,4,6-13C5 ]hexanoic acid, M5PFHxA
InChI formel InChI=1S/C6HF11O2/c7-2(8,1(18)19)3(9,10)4(11,12)5(13,14)6(15,16)17/h(H,18,19)/i1+1,2+1,3+1,4+1,6+1
LEDER O[13C](=O)[13C](F)(F)[13C](F)(F)[13C](F)(F)C(F)(F)[13C](F)(F)F
IUPAC-namn 2,2,3,3,4,4,5,5,6,6,6-undecafluoro(1,2,3,4,6-13C5)hexanoic acid
Molekylvikt (g/mol) 319.017
Formel vikt 318.997 g/mol