Läs mer
(Perfluorohexyl)acetic Acid, TRC
Produktkod. 30137815
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137815 500 mg
30010572 1 g
30004044 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137815 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP286325500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53826-12-3
Molekylformel C8 H3 F13 O2
Molekylvikt (g/mol) 378.09
Synonym 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctanoic acid, (Perfluorohexyl)acetic acid, 2H,2H-Perfluorooctanoic acid, 6:2 Fluorotelomer carboxylic acid
IUPAC-namn 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctanoic acid
LEDER OC(=O)CC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material 2H,2H-Perfluorooctanoic acid
Anteckningar om renhetsgrad GC
Kvantitet 500 mg
InChI formel InChI=1S/C8H3F13O2/c9-3(10,1-2(22)23)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h1H2,(H,22,23)
Formel vikt 377.9925
Procent renhet >95%