missing translation for 'onlineSavingsMsg'
Learn More

Perfluorohexyl bromide, 97%, Thermo Scientific Chemicals

Catalog Number p-7077789
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
335-56-8
C6BrF13
398.95
JTYRBFORUCBNHJ-UHFFFAOYSA-N
1-bromoperfluorohexane, perfluorohexyl bromide, perfluorohexylbromide, tridecafluorohexyl bromide, 1-bromo-tridecafluorohexane, bromoperfluorohexane, acmc-1cmhf, 1-bromotridecafluorohexane, tridecafluoro-1-bromohexane
92755
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane
C(C(C(C(F)(F)Br)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
This item is not returnable. View return policy
335-56-8
C6BrF13
398.95
JTYRBFORUCBNHJ-UHFFFAOYSA-N
1-bromoperfluorohexane, perfluorohexyl bromide, perfluorohexylbromide, tridecafluorohexyl bromide, 1-bromo-tridecafluorohexane, bromoperfluorohexane, acmc-1cmhf, 1-bromotridecafluorohexane, tridecafluoro-1-bromohexane
92755
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane
C(C(C(C(F)(F)Br)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.8710g/mL
Boiling Point 98.0°C to 100.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula F3C(CF2)4CF2Br
Quantity 5 g
Specific Gravity 1.871
Formula Weight 398.95
Percent Purity 97%
Chemical Name or Material Perfluorohexyl bromide
compliance-icons
Product Identifier
  • Perfluorohexyl bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'