Läs mer
Perfluorohexylphosphonic Acid 4-Methylbenzamine, TRC
Produktkod. 30419940
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30419940 500 mg
1 options
Produktkod. 30419940 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP286520500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluorohexylphosphonic Acid 4-Methylbenzamine
CAS 1263361-02-9
Molekylformel C7H9N • C6H2F13O3P
Kvantitet 500 mg
Synonym P-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)phosphonic Acid compd. with 4-Methylbenzenamine
InChI formel InChI=1S/C7H9N.C6H2F13O3P/c1-6-2-4-7(8)5-3-6;7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)23(20,21)22/h2-5H,8H2,1H3;(H2,20,21,22)
LEDER FC(F)(C(F)(F)F)C(F)(C(F)(C(F)(C(F)(P(O)(O)=O)F)F)F)F.NC1=CC=C(C=C1)C
IUPAC-namn p-toluidine (perfluorohexyl)phosphonate
Molekylvikt (g/mol) 507.19
Formel vikt 507.02