Läs mer
Perfluorononanoic acid 13C5 (1,2,3,4,5-13C5) 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Produktkod. 30322720
Change view
Kvantitet:
1 mL
Produktkod. Kvantitet
30322720 1 mL
1 options
Produktkod. 30322720 Leverantör Dr. Ehrenstorfer Leverantörsnummer DREA15987002MW50
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluorononanoic Acid 13C5 (1,2,3,4,5-13C5)
CAS 960315-49-5
Lösningsmedel eller matris Methanol/Water
Molekylformel 13C5 C4 H F17 O2
Kvantitet 1 mL
Synonym 13C5-PFNA, Perfluorononanoic Acid 13C5 (1,2,3,4,5-13C5), 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Heptadecafluorononanoic-1,2,3,4,5-13C5 acid (ACI), DTXSID 70894099, Perfluoro-n-[1,2,3,4,5-13C5]nonanoic acid, Perfluoro-n-[1,2,3,4-13C5]nonanoic acid, Nonanoic-1,2,3,4,5-13C5 acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro- (ACI)
InChI formel InChI=1S/C9HF17O2/c10-2(11,1(27)28)3(12,13)4(14,15)5(16,17)6(18,19)7(20,21)8(22,23)9(24,25)26/h(H,27,28)/i1+1,2+1,3+1,4+1,5+1
LEDER O[13C](=O)[13C](F)(F)[13C](F)(F)[13C](F)(F)[13C](F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro(1,2,3,4,5-^{13}C_{5})nonanoic acid
Molekylvikt (g/mol) 469.039
Formel vikt 468.987 g/mol