missing translation for 'onlineSavingsMsg'
Läs mer

(Perfluorooctyl)ethyl Acrylate, TRC

Produktkod. 30409492
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30409492 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30409492 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP28621510G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 1,1,2,2H-Perfluorodecyl acrylate
CAS 27905-45-9
Molekylformel C13 H7 F17 O2
Kvantitet 10 g
Synonym Acrylic acid','3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl ester (8CI), (Perfluorooctyl)ethyl acrylate, 1,1,2,2-Tetrahydroperfluorodecyl acrylate, 1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 1H,1H,2H,2H-Perfluorodecyl acrylate, 1H,1H,2H,2H-Perfluorodecyl propenoate, 2-(Perfluorooctyl)ethyl acrylate, 2-Heptadecafluorooctylethyl acrylate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl prop-2-enoate, 8:2 Fluorotelomer acrylate, AC 800, AE 800, AF 8, Apollo PC 04389E, Cheminox FAAC, Cheminox FAAC 8, FAAC, Fluowet AC 800, Fluowet AE 800, Foralkyl AC 8, Foralkyl AC 8N, Heptadecafluoro-1,1,2,2-tetrahydrodecyl acrylate, Light Acrylate FA 108, R 1820, R 1820 (ester), V 17F, V-17V, Viscoat 17F, Viscoat 17V, Zonyl TA-N
InChI formel InChI=1S/C13H7F17O2/c1-2-5(31)32-4-3-6(14,15)7(16,17)8(18,19)9(20,21)10(22,23)11(24,25)12(26,27)13(28,29)30/h2H,1,3-4H2
LEDER FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C
IUPAC-namn 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl prop-2-enoate
Molekylvikt (g/mol) 518.17
Formel vikt 518.0175
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.