Läs mer
(Perfluorooctyl)ethyl Acrylate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1,1,2,2H-Perfluorodecyl acrylate |
| CAS | 27905-45-9 |
| Molekylformel | C13 H7 F17 O2 |
| Kvantitet | 100 g |
| Synonym | Acrylic acid','3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl ester (8CI), (Perfluorooctyl)ethyl acrylate, 1,1,2,2-Tetrahydroperfluorodecyl acrylate, 1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 1H,1H,2H,2H-Perfluorodecyl acrylate, 1H,1H,2H,2H-Perfluorodecyl propenoate, 2-(Perfluorooctyl)ethyl acrylate, 2-Heptadecafluorooctylethyl acrylate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl prop-2-enoate, 8:2 Fluorotelomer acrylate, AC 800, AE 800, AF 8, Apollo PC 04389E, Cheminox FAAC, Cheminox FAAC 8, FAAC, Fluowet AC 800, Fluowet AE 800, Foralkyl AC 8, Foralkyl AC 8N, Heptadecafluoro-1,1,2,2-tetrahydrodecyl acrylate, Light Acrylate FA 108, R 1820, R 1820 (ester), V 17F, V-17V, Viscoat 17F, Viscoat 17V, Zonyl TA-N |
| InChI formel | InChI=1S/C13H7F17O2/c1-2-5(31)32-4-3-6(14,15)7(16,17)8(18,19)9(20,21)10(22,23)11(24,25)12(26,27)13(28,29)30/h2H,1,3-4H2 |
| LEDER | FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C |
| IUPAC-namn | 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl prop-2-enoate |
| Molekylvikt (g/mol) | 518.17 |
| Formel vikt | 518.0175 |