Läs mer
Perfluorooctylphosphoric Acid (>85%), TRC
Produktkod. 30411498
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30411498 100 mg
1 options
Produktkod. 30411498 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP270620100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluorooctylphosphoric Acid
CAS 40143-78-0
Anteckningar om renhetsgrad HPLC
Molekylformel C8 H2 F17 O3 P
Kvantitet 100 mg
Synonym Perfluorooctylphosphonic Acid
InChI formel InChI=1S/C8H2F17O3P/c9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)29(26,27)28/h(H2,26,27,28)
LEDER OP(=O)(O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctylphosphonic acid
Molekylvikt (g/mol) 500.0466
Formel vikt 499.947