Läs mer
Perfluoropropanesulfonic Acid Sodium, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Perfluoropropanesulfonic Acid Sodium Salt |
| CAS | 359868-82-9 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C3F7NaO3S |
| Kvantitet | 250 mg |
| Synonym | 1-Propanesulfonic acid','1,1,2,2,3,3,3-heptafluoro-','sodium salt (1:1) (ACI), 1-Propanesulfonic acid','1,1,2,2,3,3,3-heptafluoro-','sodium salt (9CI), Sodium perfluoropropanesulfonate, Perfluoropropanesulfonic acid sodium salt, Heptafluoropropane-1-sulfonic acid sodium salt, Heptafluoropropane-1-sulfonic Acid Sodium Salt, 1-Perfluoropropanesulfonic Acid Sodium Salt, 1,1,2,2,3,3,3-Heptafluoro-1-propanesulfonic Acid Sodium Salt, Heptafluoro-1-propanesulfonic Acid Sodium Salt, 1-Propanesulfonic acid','1,1,2,2,3,3,3-heptafluoro-','sodium salt (1:1) (ACI), 1-Propanesulfonic acid','1,1,2,2,3,3,3-heptafluoro-','sodium salt (9CI), Sodium perfluoropropanesulfonate |
| InChI formel | InChI=1S/C3HF7O3S.Na/c4-1(5,2(6,7)8)3(9,10)14(11,12)13;/h(H,11,12,13);/q;+1/p-1 |
| LEDER | [Na+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC-namn | sodium;1,1,2,2,3,3,3-heptafluoropropane-1-sulfonate |
| Molekylvikt (g/mol) | 272.07 |
| Visa mer |