Läs mer
Perfluoroundecanoic acid 13C2 (1,2-13C2) 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Produktkod. 30320456
Change view
Kvantitet:
1 mL
Produktkod. Kvantitet
30320456 1 mL
1 options
Produktkod. 30320456 Leverantör Dr. Ehrenstorfer Leverantörsnummer DREA15989002MW50
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluoroundecanoic Acid 13C2 (1,2-13C2)
CAS 960315-51-9
Lösningsmedel eller matris Methanol/Water
Molekylformel 13C2 C9 H F21 O2
Kvantitet 1 mL
Synonym 13C2-PFHxA, Perfluoro-n-[1,2-13C2]undecanoic Acid, Perfluoroundecanoic Acid 13C2 (1,2-13C2), 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heneicosafluoroundecanoic-1,2-13C2 acid (ACI), Perfluoro-n-[1,2-13C2]undecanoic acid, Undecanoic-1,2-13C2 acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heneico
InChI formel InChI=1S/C11HF21O2/c12-2(13,1(33)34)3(14,15)4(16,17)5(18,19)6(20,21)7(22,23)8(24,25)9(26,27)10(28,29)11(30,31)32/h(H,33,34)/i1+1,2+1
LEDER O[13C](=O)[13C](F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-henicosafluoro(1,2-^{13}C_{2})undecanoic acid
Molekylvikt (g/mol) 566.076
Formel vikt 565.971 g/mol