Läs mer
Perhexiline maleate, MedChemExpress
Produktkod. 16421108
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16421108 5 mg
16418698 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16421108 Leverantör Medchem Express Leverantörsnummer HYB1334A
MedChemExpress Perhexiline maleate is a potent carnitine palmitoyltransferase 1 (CPT 1) inhibitor with IC50s of 77 and 148 μM for rat heart and liver CPT 1, respectively.
Perhexiline maleate is a potent carnitine palmitoyltransferase 1 (CPT 1) inhibitor with IC50s of 77 and 148 μM for rat heart and liver CPT 1, respectively.
Specifikationer
Kemiskt namn eller material Perhexiline maleate
CAS 6724-53-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C23H39NO4
Kvantitet 5 mg
Löslighetsinformation Ethanol : 20 mg/mL (50.82 mM; Need ultrasonic) ∣DMSO : 3.57 mg/mL (9.07 mM; ultrasonic and warming and heat to 60°C)
LEDER O=C(O)/C=C\C(O)=O.C1(CC(C2CCCCC2)C3CCCCC3)NCCCC1
Molekylvikt (g/mol) 393.56
Formel vikt 393.56
Procent renhet 95.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death