Läs mer
Perhydrofarnesyl Acetone, TRC
Produktkod. 30412227
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30412227 50 mg
1 options
Produktkod. 30412227 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP22755050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hexahydrofarnesyl acetone
CAS 502-69-2
Molekylformel C18 H36 O
Kvantitet 50 mg
Synonym 6,10,14-Trimethyl-2-pentadecanone (ACI), (±)-Phytone, 6,10,14-Trimethylpentadecan-2-one, Hexahydrofarnesyl acetone, Perhydrofarnesyl acetone
InChI formel InChI=1S/C18H36O/c1-15(2)9-6-10-16(3)11-7-12-17(4)13-8-14-18(5)19/h15-17H,6-14H2,1-5H3
LEDER CC(C)CCCC(C)CCCC(C)CCCC(=O)C
IUPAC-namn 6,10,14-trimethylpentadecan-2-one
Molekylvikt (g/mol) 268.48
Formel vikt 268.2766