Läs mer
Perifosine, MedChemExpress
Produktkod. 16450598
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16450598 5 mg -
16434057 50 mg 50mg
16408148 10 mg -
16467896 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16450598 Leverantör Medchem Express Leverantörsnummer HY509095mg
MedChemExpress Perifosine is an oral Akt inhibitor which inhibits proliferation of different tumor cell lines with IC50s of 0.6-8.9 μM.
Perifosine is an oral Akt inhibitor which inhibits proliferation of different tumor cell lines with IC50s of 0.6-8.9 μM.
Specifikationer
Kemiskt namn eller material Perifosine
Fysisk form Solid
CAS 157716-52-4
Färg Off-White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C25H52NO4P
För användning med (applikation) Cancer-Kinase/protease
Synonym KRX-0401 NSC 639966 D21266
Löslighetsinformation H2O : ≥ 153.33 mg/mL (332.13 mM) ∣DMF : < 1 mg/mL (insoluble) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
LEDER [O-]P(OC1CC[N+](C)(CC1)C)(OCCCCCCCCCCCCCCCCCC)=O
Molekylvikt (g/mol) 461.66
Formel vikt 461.66
Procent renhet 98.0%