Läs mer
(-)-Perillyl Alcohol, TRC
Produktkod. 30179669
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179669 1 g
30179666 5 g
30179348 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179669 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2874351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18457-55-1
Molekylformel C10H16O
Molekylvikt (g/mol) 152.23
Synonym (4S)-4-(1-Methylethenyl)-1-cyclohexene-1-methanol, (-)-(S)-Perillyl Alcohol, (-)-Perillic Alcohol, (-)-Perillyl Alcohol, (-)-p-Mentha-1,8-dien-7-ol, (S)-(-)-(4-Isopropenyl-1-cyclohexenyl)methanol, (S)-(-)-Perilla Alcohol, (S)-(-)-Perillyl Alcohol, (S)-Perillic Alcohol, (S)-Perillyl Alcohol, Alda-351, Perycorolle, [(4S)-4-(Prop-1-en-2-yl)cyclohex-1-en-1-yl]methanol, [(4S)-4-Prop-1-en-2-ylcyclohexen-1-yl]methanol, l-Perillyl Alcohol;
IUPAC-namn [(4S)-4-prop-1-en-2-ylcyclohexen-1-yl]methanol
LEDER CC(=C)[C@H]1CCC(=CC1)CO
Specifikationer
Kemiskt namn eller material (-)-Perillyl Alcohol
Kvantitet 1 g
InChI formel InChI=1S/C10H16O/c1-8(2)10-5-3-9(7-11)4-6-10/h3,10-11H,1,4-7H2,2H3/t10-/m1/s1
Formel vikt 152.1201