missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5391469
Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
10450-60-9
H5IO6
227.94
MFCD00011344
TWLXDPFBEPBAQB-UHFFFAOYSA-N
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
25289
CHEBI:29150
O[I](O)(O)(O)(O)=O
This item is not returnable. View return policy
10450-60-9
H5IO6
227.94
MFCD00011344
TWLXDPFBEPBAQB-UHFFFAOYSA-N
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
25289
CHEBI:29150
O[I](O)(O)(O)(O)=O

High quality chemicals for laboratory and specialized industrial use

Packaging Wide Mouth Amber Glass Bottle
Quantity 100 g
IUPAC Name pentahydroxy(oxo)-$l^{7}-iodane
Formula Weight 227.96
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Periodic Acid
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'