missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Periostin/OSF-2 Antibody (OTI1A1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73335
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Periostin/OSF-2 Monoclonal specifically detects Periostin/OSF-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifikationer
| Periostin/OSF-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:100 | |
| MGC119510, MGC119511, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1 | |
| Mouse | |
| 93.2 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1A1 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| POSTN | |
| Human recombinant protein fragment corresponding to amino acids 1-230 of human periostin (NP_006466) produced in E.coli. | |
| Affinity Purified | |
| Cardiovascular Biology, Extracellular Matrix | |
| 10631 | |
| Human, Mouse, Rat | |
| Lyophilized |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering