missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ PERK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA515305
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PERK Polyclonal Antibody for IHC (P), Western Blot
EIF2AK3 (PERK) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK3, signaling is initiated by misfolded proteins in the ER. Defects in EIF2AK3 are the cause of Wolcott-Rallison syndrome (WRS), also known as multiple epiphyseal dysplasia with early-onset diabetes mellitus. WRS is a rare autosomal recessive disorder, characterized by permanent neonatal or early infancy insulin-dependent diabetes and, at a later age, epiphyseal dysplasia, osteoporosis, growth retardation and other multisystem manifestations, such as hepatic and renal dysfunctions, mental retardation and cardiovascular abnormalities.
Specifikationer
| PERK | |
| Polyclonal | |
| Unconjugated | |
| EIF2AK3 | |
| 2.7.11.1; AI427929; EIF2AK3; eukaryotic translation initiation factor 2 alpha kinase 3; eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; OTTHUMP00000207187; OTTHUMP00000207188; Pancreatic eIF2-alpha kinase; PEK; Perk; PRKR-like endoplasmic reticulum kinase; WRS | |
| Rabbit | |
| Size-exclusion - Dialysis, Ammonium sulfate precipitation | |
| RUO | |
| 9451 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9NZJ5 | |
| EIF2AK3 | |
| KLH conjugated synthetic peptide between 148-175 amino acids from the N-terminal region of human PERK. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering