Läs mer
GA International Permanent Autoclave-Resistant Thermal-Transfer Labels, Medium
Description
- The versatile labels can permanently adhere to a variety of surfaces such as metal, plastic, and glass
- Can resist high temperatures, ideal for multiple steam autoclave sterilization cycles, gamma irradiation (up to 50 KGy), and Ethylene oxide sterilization
- Printable with all major thermal-transfer printer brands
- Dimensions: 50.8 x 10.16 mm
- Available in various colors
Specifications
Specifications
| Autoklaveringsbar | Autoclavable |
| Typ av lim | Permanent |
| Färg | Grön |
| Avsluta | Gloss |
| Formatera | Roll |
| Material | Thermoplastic Film |
| Tryckteknik | Thermal Transfer |
| Produkttyp | Label |
| Sterilitet | Non-sterile |
| Etiketter per rad | 1 Label |
| Show More |