missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Permethrinic Acid Methyl Ester, TRC
Specifications
Specifications
| Kemiskt namn eller material | Permethrinic acid methyl ester |
| CAS | 61898-95-1 |
| Anteckningar om renhetsgrad | GC |
| Molekylformel | C9 H12 Cl2 O2 |
| Kvantitet | 50 mg |
| Synonym | Cyclopropanecarboxylic acid','3-(2,2-dichloroethenyl)-2,2-dimethyl-','methyl ester, Methoxy Permethrinate, Methyl 2,2-dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylate, Methyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate, Permethrinic acid methyl ester, Permethrin Imp. B (EP) |
| InChI formel | InChI=1S/C9H12Cl2O2/c1-9(2)5(4-6(10)11)7(9)8(12)13-3/h4-5,7H,1-3H3 |
| LEDER | COC(=O)C1C(C=C(Cl)Cl)C1(C)C |
| IUPAC-namn | methyl 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate |
| Molekylvikt (g/mol) | 223.1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?