Learn More
Perylene, 95%, BLD Pharm™
Product Code. 31065775
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31065775 25 g
31060684 5 g
31062458 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31065775 Supplier BLD Pharm Supplier No. BD14734225g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 198-55-0
Molecular Formula C20H12
MDL Number MFCD00004142
Quantity 25 g
SMILES C12=CC=CC(C3=CC=CC4=C3C5=CC=C4)=C1C5=CC=C2
Molecular Weight (g/mol) 252.31
Percent Purity 95%
Chemical Name or Material Perylene