Learn More
Gosselin™ PET Octagonal Bottles
Provides lightweight, break-resistant storage in the laboratory. Gosselin™ PET Octagonal Bottles are manufactured from polyethylene terephthalate resin.
Brand:  Gosselin™ P1000B-51
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15143927
  • 1615.00 SEK / Pack of 48
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Graduated
  • Sterile to SAL 10-3
  • Label for traceability
  • With tamper-evident screw cap
  • Compliant with ISO 5667-3 and 19458
Specifications
Specifications
1000 mL
HDPE
31mm
92.5mm
2kg
Yes
212mm
Tamper evident single closure
EC N°1935/2004, ISO 11137
Natural
PET
Sterile
92.5mm
Natural
Octagonal