Learn More
PET112 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560046
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: WKREGKTPGQ IVSEKQLELM QDQGALEQLC HSVMEAHPQV VMDVKNRNPR AINKLIGLVR KATQSRADPV MIKEILEKK Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 73%.
Allows the formation of correctly charged Gln-tRNA(Gln) through the transamidation of misacylated Glu-tRNA(Gln) in the mitochondria. The reaction takes place in the presence of glutamine and ATP through an activated gamma-phospho-Glu-tRNA(Gln). [UniProt]Specifications
PET112 | |
Polyclonal | |
Unconjugated | |
GATB | |
9430026F02Rik; cytochrome c oxidase assembly factor PET112 homolog; cytochrome oxidase assembly factor PET112 homolog; Gatb; Glu-AdT subunit B; glutamyl-tRNA amidotransferase subunit B; glutamyl-tRNA(Gln) amidotransferase subunit B, mitochondrial; glutamyl-tRNA(Gln) amidotransferase, subunit B; HSPC199; Pet112; PET112 homolog; PET112L; probable glutamyl-tRNA(Gln) amidotransferase subunit B, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5188 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75879 | |
GATB | |
Recombinant protein corresponding to Human PET112 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only