Läs mer
Petesicatib, MedChemExpress
Produktkod. 16490597
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16490597 50 mg
16486987 5 mg
16485196 100 mg
16474268 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16490597 Leverantör Medchem Express Leverantörsnummer HY109069
MedChemExpress Petesicatib is a cathepsin S inhibitor, used in research of immune diseases.
Petesicatib is a cathepsin S inhibitor, used in research of immune diseases.
Specifikationer
Kemiskt namn eller material Petesicatib
CAS 1252637-35-6
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C25H23F6N5O4S
Kvantitet 50 mg
Löslighetsinformation DMSO : 125 mg/mL (207.11 mM; Need ultrasonic)
LEDER O=C(N1[C@@H](C[C@@H](S(=O)(C2=C(C=C(C3=CN(C)N=C3)C=C2)C(F)(F)F)=O)C1)C(NC4(CC4)C#N)=O)C5(CC5)C(F)(F)F
Molekylvikt (g/mol) 603.54
Formel vikt 603.54
Procent renhet 99.71%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation