Learn More
Systec™ Petri Dish Stacker Carousel
Systec™ Petri Dish Stacker Carousel is suitable for containing 90mm petri dishes in a plate pourer array.
Brand: Systec™
Additional Details : Weight : 7.00000kg
Description
- For 90mm petri dishes
- For plate pourer array
Specifications
Petri dishes, 90 mm |