missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether 35/60, ACS grade, Alfa Aesar™

Maximum level of impurities: Color (APHA): 10, Boiling range 35-60°C, Evaporation residue: 0.001%, Acidity P.T.

Brand:  Alfa Aesar 042085.M1

Additional Details : CAS Number : 8032-32-4 Weight : 1.00000kg Transport : UN number : 1268 Chem class : 1 Pack group : I

Quantity 4 x 1L

 View more versions of this product

Product Code. 15471777

  • 1263.04 SEK / 1 L


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Specifications

Chemical Identifiers

Petroleum ether 35/60
ACS Grade
MFCD00081849
8032-32-4
Liquid
Ligroin

Specifications

Petroleum ether 35/60
ACS Grade
Liquid
Ligroin
Petroleum-like
Clear colorless
8032-32-4
Colorless liquid
MFCD00081849
250°C (482°F)
350 hPa (263mm Hg) at 20°C
4 x 1L
Videos
SDS