Läs mer
Petromyzonol 24-Sulfate Sodium Salt, TRC
Produktkod. 30417981
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417981 1 mg
30415635 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30417981 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2935261MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1379583-33-1
Molekylformel C24 H41 O7 S . Na
Molekylvikt (g/mol) 496.63
IUPAC-namn sodium;[(4R)-4-[(3R,5R,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentyl] sulfate
LEDER C[C@@]12[C@@]3([C@]([C@]4([C@](C)([C@@H](O)C3)[C@@]([C@@H](CCCOS(=O)(=O)O)C)(CC4)[H])[H])([C@H](O)C[C@]1(C[C@H](O)CC2)[H])[H])[H].[Na]
Specifikationer
Kemiskt namn eller material Petromyzonol-24-Sulfate, Sodium Salt
Kvantitet 1 mg
InChI formel InChI=1S/C24H42O7S.Na/c1-14(5-4-10-31-32(28,29)30)17-6-7-18-22-19(13-21(27)24(17,18)3)23(2)9-8-16(25)11-15(23)12-20(22)26;/h14-22,25-27H,4-13H2,1-3H3,(H,28,29,30);/t14-,15-,16-,17-,18+,19+,20-,21+,22+,23+,24-;/m1./s1
Formel vikt 496.2471