Läs mer
Tocris Bioscience™ PF 04449913 maleate
Beskrivning
PF 04449913 maleate is a potent Smo antagonist (IC50 = 5 nM). Attenuates the leukemia-initiation potential of AML cells in a serial transplantation mouse model. Also eliminates self-propagation capacity of AML cells.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1-((2R,4R)-2-(1H-Benzimidazol-2-yl)-1-methylpiperidin-4-yl)-3-(4-cyanophenyl)urea maleate salt |
| CAS | 2030410-25-2 |
| Synonym | glasdegib, PF-913 |
| Kvantitet | 5 mg |
| Molekylformel | C21H22N6O.C4H4O4 |
| Renhet | 98% |