Learn More
Tocris Bioscience™ PF 04628935
Description
PF 04628935 is a potent ghrelin receptor inverse agonist (IC50 = 4.6 nM). Orally bioavailable and exhibits reasonable brain penetration.
Specifications
Specifications
| Kemiskt namn eller material | 1-[2-[[2-Chloro-4-(2H-1,2,3-triazol-2-yl)phenyl]methyl]-2,7-diazaspiro[3.5]non-7-yl]-2-(2-methylimidazo[2,1-b]thiazol-6-yl)ethanone |
| CAS | 1383719-97-8 |
| Analysprocentintervall | >98% |
| Kvantitet | 25 mg |
| Molekylformel | C24H26ClN7OS |
| Formel vikt | Observed MW: 496.03 |