Läs mer
PF-04979064, MedChemExpress
Produktkod. 16485727
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
Förpackningsstorlek:
10mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16485727 5 mg 5mg
16479556 50 mg -
16472988 10 mg 10mg
16432576 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16485727 Leverantör Medchem Express Leverantörsnummer HY100398
MedChemExpress PF-04979064 is a potent and selective PI3K/mTOR dual kinase inhibitor with Kis of 0.13 nM and 1.42 nM for PI3Kα and mTOR, respectively.
PF-04979064 is a potent and selective PI3K/mTOR dual kinase inhibitor with Kis of 0.13 nM and 1.42 nM for PI3Kα and mTOR, respectively.
Specifikationer
Kemiskt namn eller material PF-04979064
CAS 1220699-06-8
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C24H26N6O3
Kvantitet 5 mg
Löslighetsinformation DMSO : 10 mg/mL (22.40 mM; Need ultrasonic)
LEDER O=C(N1C2CCN(C([C@@H](O)C)=O)CC2)N(C)C3=C1C4=NC(C5=CC=C(C)N=C5)=CC=C4N=C3
Molekylvikt (g/mol) 446.5
Formel vikt 446.5
Procent renhet 99.83%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease