Läs mer
PF 05180999, Tocris Bioscience™
Potent and selective PDE2A inhibitor
Brand: Tocris Bioscience 6405/5
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PF 05180999 is a potent and selective PDE2A inhibitor (IC50 = 1 nM). Exhibits 2000-fold selectivity for PDE2A over PDE10A. Brain penetrant.Specifikationer
| 4-(1-Azetidinyl)-7-methyl-5-[1-methyl-5-[5-(trifluoromethyl)-2-pyridinyl]-1H-pyrazol-4-yl]-imidazo[5,1-f][1,2,4]triazine | |
| 1394033-54-5 | |
| >98% | |
| 5 mg |