missing translation for 'onlineSavingsMsg'
Läs mer

PF-06305591, MedChemExpress

Produktkod. 16437557
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16437557 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16437557 Leverantör Medchem Express Leverantörsnummer HY114301

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress PF-06305591 is a potent and highly selective voltage gated sodium channel NaV1.8 blocker, with an IC50 of 15 nM. An excellent preclinical in vitro ADME and safety profile.

PF-06305591 is a potent and highly selective voltage gated sodium channel NaV1.8 blocker, with an IC50 of 15 nM. An excellent preclinical in vitro ADME and safety profile.

Specifikationer

Kemiskt namn eller material PF-06305591
Fysisk form Powder
CAS 1449473-97-5
Färg Off-White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C15H22N4O
För användning med (applikation) Neuroscience-Neuromodulation
Löslighetsinformation DMSO : 50 mg/mL (182.24 mM; Need ultrasonic)
LEDER CC(C)(C)C1=CC=C(NC([C@H]([C@@H](C)C(N)=O)N)=N2)C2=C1
Molekylvikt (g/mol) 274.36
Formel vikt 274.36
Procent renhet 99.0%
Kvalitet Research
Visa mer Visa mindre

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.