Läs mer
Tocris Bioscience™ PF 06465469
GREENER_CHOICE
Produktkod. 30001571
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001571 50 mg
30001555 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001571 Leverantör Tocris Bioscience™ Leverantörsnummer 4710/50
Potent ITK inhibitor; also inhibits BTK
Potent inhibitor of interleukin-2 inducible T cell kinase (ITK) (IC50 = 2 nM). Also exhibits inhibitory activity against Bruton's tyrosine kinase (BTK) (IC50 = 2 nM). Displays nanomolar potencies in a cell-based IP1 assay and human whole blood assay (IC50 values are 31 and 48 nM respectively).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1407966-77-1
Molekylformel C30H33N7O2
Specifikationer
Kemiskt namn eller material PF 06465469
Kvantitet 50 mg
Formel vikt 523.63
Renhet >95%
Molekylvikt (g/mol) 523.641